3,5-Dibromo-1-(trimethylsilyl)benzene ≥97.0%

Supplier: TCI America

D2724-25G D2724-5G
TCD2724-25GEA 1551.56 CAD
TCD2724-25G TCD2724-005G
3,5-Dibromo-1-(trimethylsilyl)benzene ≥97.0%
3,5-Dibromo-1-(trimethylsilyl)benzene

CAS Number: 17878-23-8
MDL Number: MFCD03844787
Molecular Formula: C9H12Br2Si
Molecular Weight: 308.09
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 40


Product Categories: Materials Chemistry, Dendrimer Building Blocks
Other Product Names / Synonyms: (3,5-Dibromophenyl)trimethylsilane

Formula: C₉H₁₂Br₂Si
Melting Pt: 40 °C
MDL Number: MFCD03844787
CAS Number: 17878-23-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd560a904a1d670b759833e69e797e0d5ce10f90&vid=dd560a904a1d670b759833e69e797e0d5ce10f90&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR