(±)-1,2-Decanediol ≥99.0%

Supplier: TCI America

D2720-25G D2720-100G
TCD2720-25GEA 67.53 CAD
TCD2720-25G TCD2720-100G
(±)-1,2-Decanediol ≥99.0%
(±)-1,2-Decanediol

CAS Number: 1119-86-4
MDL Number: MFCD00010739
Molecular Formula: C10H22O2
Molecular Weight: 174.28
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 168
Melting point (°C): 49
Flash Point (°C): 110


Other Product Names / Synonyms: 1,2-Dihydroxydecane

Formula: C₁₀H₂₂O₂
Boiling Pt: 168 °C
Melting Pt: 49 °C
Flash Pt: 110 °C
MDL Number: MFCD00010739
CAS Number: 1119-86-4
EINECS: 214-288-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a58bf929f446c50fc0d1b38a2ae0ea752426e151&vid=a58bf929f446c50fc0d1b38a2ae0ea752426e151&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR