erythro-5,6-Dodecanediol ≥98.0%
Supplier: TCI America
Synonyms:
erythro-5,6-Dihydroxydodecane
D2719-1G
D2719-100MG
TCD2719-001GEA
3885.22
CAD
TCD2719-001G
TCD2719-100MG
erythro-5,6-Dodecanediol ≥98.0%
erythro-5,6-Dodecanediol
CAS Number: 70859-32-4
MDL Number: MFCD02093467
Molecular Formula: C12H26O2
Molecular Weight: 202.34
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Other Product Names / Synonyms: erythro-5,6-Dihydroxydodecane
Formula:
C₁₂H₂₆O₂ MW: 202.34 g/mol |
MDL Number:
MFCD02093467 CAS Number: 70859-32-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...