4,4'-Dihydroxytetraphenylmethane ≥98.0%
Supplier: TCI America
Synonyms:
(Diphenyl)bis(4-hydroxyphenyl)methane, Bis(4-hydroxyphenyl)diphenylmethane
D2711-25G
D2711-5G
TCD2711-25GEA
314.42
CAD
TCD2711-25G
TCD2711-005G
4,4'-Dihydroxytetraphenylmethane ≥98.0%
4,4'-Dihydroxytetraphenylmethane
CAS Number: 1844-01-5
MDL Number: MFCD00045785
Molecular Formula: C25H20O2
Molecular Weight: 352.43
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Product Categories: Materials Chemistry, Monomers, Diphenylmethanes (for High-Performance Polymer Research)
Other Product Names / Synonyms: Bis(4-hydroxyphenyl)diphenylmethane, (Diphenyl)bis(4-hydroxyphenyl)methane
Formula:
C₂₅H₂₀O₂ MW: 352.43 g/mol |
MDL Number:
MFCD00045785 CAS Number: 1844-01-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...