3,5-Diamino-2,4,6-trimethylbenzenesulfonic acid ≥93.0%
Supplier: TCI America
Synonyms:
2,6-Diaminomesitylene-4-sulphonic acid, 2,6-Diaminomesitylene-4-sulfonic acid, 2,4,6-Trimethyl-3,5-diaminobenzenesulfonic acid, 2,4,6-Trimethyl-3,5-diaminobenzenesulphonic acid
D2709-5G
D2709-25G
TCD2709-005GEA
25.72
CAD
TCD2709-005G
TCD2709-025G
3,5-Diamino-2,4,6-trimethylbenzenesulfonic acid ≥93.0%
3,5-Diamino-2,4,6-trimethylbenzenesulfonic acid
CAS Number: 32432-55-6
MDL Number: MFCD00043827
Molecular Formula: C9H14N2O3S
Molecular Weight: 230.28
Purity/Analysis Method: >93.0% (T)
Form: Crystal
Other Product Names / Synonyms: 2,6-Diaminomesitylene-4-sulfonic Acid, 2,4,6-Trimethyl-3,5-diaminobenzenesulfonic Acid
Formula:
C₉H₁₄N₂O₃S MW: 230.28 g/mol |
MDL Number:
MFCD00043827 CAS Number: 32432-55-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...