2,4-Dimethoxy-4'-hydroxybenzophenone ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (2,4-Dimethoxyphenyl)(4-hydroxyphenyl)methanone

D2707-5G D2707-25G
TCD2707-005GEA 107.44 CAD
TCD2707-005G TCD2707-025G
2,4-Dimethoxy-4'-hydroxybenzophenone ≥98.0% (by titrimetric analysis)
2,4-Dimethoxy-4'-hydroxybenzophenone

CAS Number: 41351-30-8
MDL Number: MFCD00137850
Molecular Formula: C15H14O4
Molecular Weight: 258.27
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 153


Product Categories: Synthetic Organic Chemistry, Linkers (Combinatorial Chemistry)
Other Product Names / Synonyms: 4'-Hydroxy-2,4-dimethoxybenzophenone

Formula: C₁₅H₁₄O₄
MW: 258.27 g/mol
Melting Pt: 153 °C
MDL Number: MFCD00137850
CAS Number: 41351-30-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5040f02015be589955ff6b870efb39179990f385&vid=5040f02015be589955ff6b870efb39179990f385&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR