Diisodecyl adipate
Supplier: TCI America
A0164-500ML
A0164-25ML
TCA0164-500MLEA
85.32
CAD
TCA0164-500ML
TCA0164-025ML
Diisodecyl adipate
Diisodecyl adipate
CAS Number: 27178-16-1
MDL Number: MFCD00048392
Molecular Weight: 426.68
Form: Clear Liquid
Boiling point (°C): 240
Flash Point (°C): 229
Specific Gravity (20/20): 0.92
Other Product Names / Synonyms: Adipic Acid Diisodecyl Ester
Boiling Pt:
240 °C Density: 0.92 (20 °C) Flash Pt: 229 °C |
MDL Number:
MFCD00048392 CAS Number: 27178-16-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...