(-)-Dipivaloyl-L-tartaric acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
D2686-25G
D2686-5G
TCD2686-25GEA
335.59
CAD
TCD2686-25G
TCD2686-005G
(-)-Dipivaloyl-L-tartaric acid ≥98.0% (by titrimetric analysis)
(-)-Dipivaloyl-L-tartaric acid
CAS Number: 65259-81-6
MDL Number: MFCD00066535
Molecular Formula: C14H22O8
Molecular Weight: 318.32
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 150
Specific rotation [a]20/D: -24 deg (C=2, Dioxane)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (-)-Bis(2,2-dimethylpropionyl)-L-tartaric Acid
Formula:
C₁₄H₂₂O₈ Melting Pt: 150 °C |
MDL Number:
MFCD00066535 CAS Number: 65259-81-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...