3,5-Di-tert-butylbenzyl alcohol ≥98.0%

Supplier: TCI America

D2652-1G D2652-5G
TCD2652-001GEA 112.34 CAD
TCD2652-001G TCD2652-5G
3,5-Di-tert-butylbenzyl alcohol ≥98.0%
3,5-Di-tert-butylbenzyl alcohol

CAS Number: 77387-57-6
MDL Number: MFCD03844785
Molecular Formula: C15H24O
Molecular Weight: 220.36
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 61

Formula: C₁₅H₂₄O
Melting Pt: 61 °C
MDL Number: MFCD03844785
CAS Number: 77387-57-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85169dec878e7088ab9ac606d0f47da3866edecb&vid=85169dec878e7088ab9ac606d0f47da3866edecb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR