3,6-Dioxa-1,8-octanedithiol ≥97.0%

Supplier: TCI America

Synonyms: Ethylene Glycol Bis(2-mercaptoethyl) Ether, 1,8-Dimercapto-3,6-dioxaoctane, 1,2-Bis(2-mercaptoethoxy)ethane, 2,2′-(Ethylenedioxy)diethanethiol

D2649-25G D2649-500G D2649-100G
TCD2649-025GEA 52.9 CAD
TCD2649-025G TCD2649-500G TCD2649-100G
3,6-Dioxa-1,8-octanedithiol ≥97.0%
3,6-Dioxa-1,8-octanedithiol

CAS Number: 14970-87-7
MDL Number: MFCD00015873
Molecular Formula: C6H14O2S2
Molecular Weight: 182.30
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Boiling point (°C): 86
Flash Point (°C): 132
Specific Gravity (20/20): 1.12


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,2-Bis(2-mercaptoethoxy)ethane, 1,8-Dimercapto-3,6-dioxaoctane, Ethylene Glycol Bis(2-mercaptoethyl) Ether

Formula: C₆H₁₄O₂S₂
MW: 182.30 g/mol
Boiling Pt: 86 °C
Density: 1.12 (20 °C)
Flash Pt: 132 °C
MDL Number: MFCD00015873
CAS Number: 14970-87-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7bd1aca90f4ac9aaf79ad7c074555af5011b5822&vid=7bd1aca90f4ac9aaf79ad7c074555af5011b5822&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR