(-)-Diacetyl-D-tartaric anhydride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
D2645-5G
D2645-25G
TCD2645-005GEA
170.37
CAD
TCD2645-005G
TCD2645-025G
(-)-Diacetyl-D-tartaric anhydride ≥98.0% (by titrimetric analysis)
(-)-Diacetyl-D-tartaric anhydride
CAS Number: 70728-23-3
MDL Number: MFCD02093461
Molecular Formula: C8H8O7
Molecular Weight: 216.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 134
Specific rotation [a]20/D: -96 deg (C=0.5, CHCl3)
Storage Temperature: <0°C
Formula:
C₈H₈O₇ Melting Pt: 134 °C |
MDL Number:
MFCD02093461 CAS Number: 70728-23-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...