N,N-Dimethylsulfamide ≥98.0% (by total nitrogen basis)

Supplier: TCI America

D2640-5G D2640-1G
TCD2640-5GEA 583.02 CAD
TCD2640-5G TCD2640-001G
N,N-Dimethylsulfamide ≥98.0% (by total nitrogen basis)
N,N-Dimethylsulfamide

CAS Number: 3984-14-3
MDL Number: MFCD01861286
Molecular Formula: C2H8N2O2S
Molecular Weight: 124.16
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 97


Other Product Names / Synonyms: N,N-Dimethylsulfamoylamide

Formula: C₂H₈N₂O₂S
Melting Pt: 97 °C
MDL Number: MFCD01861286
CAS Number: 3984-14-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da2568f11ac3e70903102b6e054ec01ead1a3bce&vid=da2568f11ac3e70903102b6e054ec01ead1a3bce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR