2,5-Diamino-4,6-dihydroxypyrimidine hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: 2,5-Diaminopyrimidine-4,6-diol Hydrochloride, 2,5-Diamino-6-hydroxy-4(1H)-pyrimidinone Hydrochloride

D2634-25G D2634-5G
TCD2634-025GEA 469.42 CAD
TCD2634-025G TCD2634-005G
2,5-Diamino-4,6-dihydroxypyrimidine hydrochloride ≥98.0%
2,5-Diamino-4,6-dihydroxypyrimidine hydrochloride

CAS Number: 56830-58-1
MDL Number: MFCD00082893
Molecular Formula: C4H6N4O2
Molecular Weight: 178.58
Purity/Analysis Method: >98.0% (T,HPLC)
Form: Crystal


Other Product Names / Synonyms: 2,5-Diamino-6-hydroxy-4(1H)-pyrimidinone Hydrochloride, 2,5-Diaminopyrimidine-4,6-diol Hydrochloride

Formula: C₄H₆N₄O₂
MW: 178.58 g/mol
MDL Number: MFCD00082893
CAS Number: 56830-58-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ef11aa5457a93597fd8b4832a7f93d0ab88fc60&vid=6ef11aa5457a93597fd8b4832a7f93d0ab88fc60&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T,HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR