2-Amino-3,5-dibromobenzaldehyde ≥98.0%

Supplier: TCI America
Warning

D2625-5G D2625-25G
TCD2625-005GEA 37.92 CAD
TCD2625-005G TCD2625-025G
2-Amino-3,5-dibromobenzaldehyde ≥98.0%
2-Amino-3,5-dibromobenzaldehyde

CAS Number: 50910-55-9
MDL Number: MFCD00671100
Molecular Formula: C7H5Br2NO
Molecular Weight: 278.93
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 137


Other Product Names / Synonyms: 2-Amino-3,5-dibromobenzaldehyde

Formula: C₇H₅Br₂NO
Boiling Pt: 331 °C (1013 hPa)
Melting Pt: 137 °C
MDL Number: MFCD00671100
CAS Number: 50910-55-9
EINECS: 256-841-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4cce03c5651a0e220495b08bd560cbee6fb5437&vid=77c0b691d96751f751c8781967d82334a729d321&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR