Diethylene glycol monovinyl ether ≥96.0% stabilized

Supplier: TCI America

Synonyms: Vinyl Carbitol, 2-(2-Vinyloxyethoxy)ethanol, Di(ethylene glycol) vinyl ether, 2-(2-(Vinyloxy)ethoxy)ethanol

D2623-25ML D2623-500ML D2623-100ML
TCD2623-025MLEA 83.74 CAD
TCD2623-025ML TCD2623-500ML TCD2623-100ML
Diethylene glycol monovinyl ether ≥96.0% stabilized
Diethylene glycol monovinyl ether

(stabilized with KOH)
CAS Number: 929-37-3
MDL Number: MFCD00192027
Molecular Formula: C6H12O3
Molecular Weight: 132.16
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 96
Flash Point (°C): 82
Specific Gravity (20/20): 1.03


Product Categories: Bioscience, Monomers for Synthetic Polymers (for DDS)
Other Product Names / Synonyms: Vinyl Carbitol, 2-(2-Vinyloxyethoxy)ethanol

Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 96 °C
Density: 1.03 (20 °C)
Flash Pt: 82 °C
MDL Number: MFCD00192027
CAS Number: 929-37-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c23c6ea908f07f530095ddfddcc0a3f6c0c07ca1&vid=c23c6ea908f07f530095ddfddcc0a3f6c0c07ca1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR