4-(Hydroxymethyl)pyrocatechol ≥96.0%

Supplier: TCI America

D2620-1G D2620-5G
TCD2620-001GEA 197.5 CAD
TCD2620-001G TCD2620-005G
4-(Hydroxymethyl)pyrocatechol ≥96.0%
4-(Hydroxymethyl)pyrocatechol

CAS Number: 3897-89-0
MDL Number: MFCD00210409
Molecular Formula: C7H8O3
Molecular Weight: 140.14
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 117
Storage Temperature: <0°C


Other Product Names / Synonyms: 4-Hydroxymethylpyrocatechol, alpha,3,4-Trihydroxytoluene

Formula: C₇H₈O₃
Melting Pt: 117 °C
MDL Number: MFCD00210409
CAS Number: 3897-89-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=366273a3f89d304009e8c90e5c161d4e5f4df681&vid=366273a3f89d304009e8c90e5c161d4e5f4df681&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR