2,4-Diethyl-1,5-pentanediol (DL- and meso- mixture) ≥93.0%

Supplier: TCI America

D2619-25ML D2619-500ML
TCD2619-025MLEA 26.86 CAD
TCD2619-025ML TCD2619-500ML
2,4-Diethyl-1,5-pentanediol (DL- and meso- mixture) ≥93.0%
2,4-Diethyl-1,5-pentanediol (DL- and meso- mixture)

CAS Number: 57987-55-0
MDL Number: MFCD01861285
Molecular Formula: C9H20O2
Molecular Weight: 160.26
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Boiling point (°C): 150
Flash Point (°C): 143
Specific Gravity (20/20): 0.95

Formula: C₉H₂₀O₂
MW: 160.26 g/mol
Boiling Pt: 150 °C
Density: 0.95 (20 °C)
Flash Pt: 143 °C
MDL Number: MFCD01861285
CAS Number: 57987-55-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc3d6a426cc23754c2080e007a868c3659bb8afe&vid=cc3d6a426cc23754c2080e007a868c3659bb8afe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR