2,2'-Dinitrodibenzyl ≥98.0%
Supplier: TCI America
D2602-5G
D2602-25G
TCD2602-005GEA
121.66
CAD
TCD2602-005G
TCD2602-025G
2,2'-Dinitrodibenzyl ≥98.0%
2,2'-Dinitrodibenzyl
CAS Number: 16968-19-7
MDL Number: MFCD00024296
Molecular Formula: C14H12N2O4
Molecular Weight: 272.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 122
Other Product Names / Synonyms: 1,2-Bis(2-nitrophenyl)ethane, 2,2'-Dinitrobibenzyl
Formula:
C₁₄H₁₂N₂O₄ Melting Pt: 122 °C |
MDL Number:
MFCD00024296 CAS Number: 16968-19-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...