2,6-Dibromo-1,5-dihydroxynaphthalene ≥93.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2,6-Dibromonaphthalene-1,5-diol, 2,6-Dibromo-1,5-naphthalenediol
D2593-25G
D2593-5G
TCD2593-25GEA
1597.38
CAD
TCD2593-25G
TCD2593-005G
2,6-Dibromo-1,5-dihydroxynaphthalene ≥93.0% (by titrimetric analysis)
2,6-Dibromo-1,5-dihydroxynaphthalene
CAS Number: 84-59-3
MDL Number: MFCD00021473
Molecular Formula: C10H6Br2O2
Molecular Weight: 317.96
Purity/Analysis Method: >93.0% (T)
Form: Crystal
Other Product Names / Synonyms: 2,6-Dibromo-1,5-naphthalenediol
Formula:
C₁₀H₆Br₂O₂ MW: 317.96 g/mol |
MDL Number:
MFCD00021473 CAS Number: 84-59-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...