2,6-Dimethoxy-p-cresol ≥97.0%

Supplier: TCI America

Synonyms: 3,5-Dimethoxy-4-hydroxytoluene, 4-Methyl-2,6-dimethoxyphenol, 1,3-Dimethoxy-2-hydroxy-5-methylbenzene, 2,6-Dimethoxy-4-methylphenol

D2591-5G D2591-25G
TCD2591-005GEA 56.88 CAD
TCD2591-005G TCD2591-025G
2,6-Dimethoxy-p-cresol ≥97.0%
2,6-Dimethoxy-p-cresol

CAS Number: 6638-05-7
MDL Number: MFCD00017289
Molecular Formula: C9H12O3
Molecular Weight: 168.19
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 150
Melting point (°C): 42
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2,6-Dimethoxy-p-cresol, 4-Hydroxy-3,5-dimethoxytoluene

Formula: C₉H₁₂O₃
MW: 168.19 g/mol
Boiling Pt: 150 °C
Melting Pt: 42 °C
MDL Number: MFCD00017289
CAS Number: 6638-05-7
EINECS: 229-641-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ef4ebb787f144499e0aa219838c7cd41d4d203b&vid=0ef4ebb787f144499e0aa219838c7cd41d4d203b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR