5,6-Dihydro-1,4-dithiin-2,3-dicarboximide ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2,3-Dihydro-1,4-dithiino[2,3-c]pyrrole-5,7-dione
D2574-1G
D2574-5G
TCD2574-001GEA
156.42
CAD
TCD2574-001G
TCD2574-005G
5,6-Dihydro-1,4-dithiin-2,3-dicarboximide ≥97.0% (by titrimetric analysis)
5,6-Dihydro-1,4-dithiin-2,3-dicarboximide
CAS Number: 24519-85-5
MDL Number: MFCD01632185
Molecular Formula: C6H5NO2S2
Molecular Weight: 187.23
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 217
Other Product Names / Synonyms: 2,3-Dihydro-1,4-dithiino[2,3-c]pyrrole-5,7-dione
Formula:
C₆H₅NO₂S₂ MW: 187.23 g/mol Melting Pt: 217 °C |
MDL Number:
MFCD01632185 CAS Number: 24519-85-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...