N,N'-Bis(salicylidene)-1,2-phenylenediamine ≥96.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: N,N'-Bis(2-hydroxybenzylidene)-1,2-phenylenediamine, N,N'-Disalicylal-1,2-phenylenediamine, 2,2'-((1,2-Phenylenebis(azanylylidene))bis(methanylylidene))diphenol

D2572-5G D2572-25G
TCD2572-005GEA 151.68 CAD
TCD2572-005G TCD2572-025G
N,N'-Bis(salicylidene)-1,2-phenylenediamine ≥96.0% (by HPLC, total nitrogen)
N,N'-Bis(salicylidene)-1,2-phenylenediamine

CAS Number: 3946-91-6
MDL Number: MFCD00009995
Molecular Formula: C20H16N2O2
Molecular Weight: 316.36
Purity/Analysis Method: >96.0% (HPLC,N)
Form: Crystal
Melting point (°C): 165


Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: N,N'-Bis(2-hydroxybenzylidene)-1,2-phenylenediamine, N,N'-Disalicylal-1,2-phenylenediamine

Formula: C₂₀H₁₆N₂O₂
MW: 316.36 g/mol
Melting Pt: 165 °C
MDL Number: MFCD00009995
CAS Number: 3946-91-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b0d143d3bcfcb9800b654f7eee2ae280b4bd362&vid=8b0d143d3bcfcb9800b654f7eee2ae280b4bd362&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR