2,4'-Dihydroxydiphenyl sulfone ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-((4-Hydroxyphenyl)sulfonyl)phenol, 2-((4-Hydroxyphenyl)sulphonyl)phenol

D2567-500G D2567-25G
TCD2567-500GEA 385.2 CAD
TCD2567-500G TCD2567-025G
2,4'-Dihydroxydiphenyl sulfone ≥98.0% (by titrimetric analysis)
2,4'-Dihydroxydiphenyl sulfone

CAS Number: 5397-34-2
MDL Number: MFCD01631305
Molecular Formula: C12H10O4S
Molecular Weight: 250.27
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 184

Formula: C₁₂H₁₀O₄S
MW: 250.27 g/mol
Melting Pt: 184 °C
MDL Number: MFCD01631305
CAS Number: 5397-34-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eca2f1bb05ef566dec9a28c62605940dcd910dc8&vid=eca2f1bb05ef566dec9a28c62605940dcd910dc8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR