2,9-Dibutyl-1,10-phenanthroline ≥98.0% (by total nitrogen basis)

Supplier: TCI America

D2565-1G D2565-100MG
TCD2565-001GEA 784.94 CAD
TCD2565-001G TCD2565-100MG
2,9-Dibutyl-1,10-phenanthroline ≥98.0% (by total nitrogen basis)
2,9-Dibutyl-1,10-phenanthroline

CAS Number: 85575-93-5
MDL Number: MFCD02093459
Molecular Formula: C20H24N2
Molecular Weight: 292.43
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 58

Formula: C₂₀H₂₄N₂
Melting Pt: 58 °C
MDL Number: MFCD02093459
CAS Number: 85575-93-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c58aeea1d47e8f4461b02dcaabb94845814244d&vid=0c58aeea1d47e8f4461b02dcaabb94845814244d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR