2,6-Diphenylphenol ≥98.0%

Supplier: TCI America

Synonyms: [1,1':3',1''-Terphenyl]-2'-ol

D2563-25G D2563-5G
TCD2563-025GEA 294.1 CAD
TCD2563-025G TCD2563-005G
2,6-Diphenylphenol ≥98.0%
2,6-Diphenylphenol

CAS Number: 2432-11-3
MDL Number: MFCD00009716
Molecular Formula: C18H14O
Molecular Weight: 246.31
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 103


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 2'-Hydroxy-m-terphenyl

Formula: C₁₈H₁₄O
MW: 246.31 g/mol
Melting Pt: 103 °C
MDL Number: MFCD00009716
CAS Number: 2432-11-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a31194a03fe17152fb743915a3344c7d8135819e&vid=a31194a03fe17152fb743915a3344c7d8135819e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR