Methyl (R)-(+)-2,2-dimethyl-1,3-dioxolane-4-carboxylate ≥98.0%

Supplier: TCI America

Synonyms: (+)-2,3-O-Isopropylidene-D-glyceric Acid Methyl Ester, (R)-Methyl 2,2-dimethyl-1,3-dioxolane-4-carboxylate, (+)-Methyl 2,3-O-Isopropylidene-D-glycerate, (R)-(+)-2,2-Dimethyl-1,3-dioxolane-4-carboxylic Acid Methyl Ester

D2562-25G D2562-5G
TCD2562-025GEA 551.42 CAD
TCD2562-025G TCD2562-5G
Methyl (R)-(+)-2,2-dimethyl-1,3-dioxolane-4-carboxylate ≥98.0%
Methyl (R)-(+)-2,2-dimethyl-1,3-dioxolane-4-carboxylate

CAS Number: 52373-72-5
MDL Number: MFCD00066527
Molecular Formula: C7H12O4
Molecular Weight: 160.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 78
Flash Point (°C): 75
Specific Gravity (20/20): 1.11
Specific rotation [a]20/D: 11 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (+)-2,3-O-Isopropylidene-D-glyceric Acid Methyl Ester, (R)-(+)-2,2-Dimethyl-1,3-dioxolane-4-carboxylic Acid Methyl Ester, (+)-Methyl 2,3-O-Isopropylidene-D-glycerate

Formula: C₇H₁₂O₄
MW: 160.17 g/mol
Boiling Pt: 78 °C
Density: 1.11 (20 °C)
Flash Pt: 75 °C
MDL Number: MFCD00066527
CAS Number: 52373-72-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6eab0528fe491acade239f6ee7be2b9b990f59e3&vid=6eab0528fe491acade239f6ee7be2b9b990f59e3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR