1,2:3,4-Di-O-isopropylidene-ɑ-D-galactopyranose ≥92.0%
Supplier: TCI America
D2555-5G
D2555-25G
TCD2555-005GEA
154.84
CAD
TCD2555-005G
TCD2555-025G
1,2:3,4-Di-O-isopropylidene-ɑ-D-galactopyranose ≥92.0%
1,2:3,4-Di-O-isopropylidene-alpha-D-galactopyranose
CAS Number: 4064-06-6
MDL Number: MFCD00063225
Molecular Formula: C12H20O6
Molecular Weight: 260.29
Purity/Analysis Method: >92.0% (GC)
Form: Starch Syrup
Boiling point (°C): 133
Specific rotation [a]20/D: -58 deg (C=3, CHCl3)
Formula:
C₁₂H₂₀O₆ Boiling Pt: 133 °C |
MDL Number:
MFCD00063225 CAS Number: 4064-06-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...