(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethyl-p-toluenesulfonate ≥98.0%
Supplier: TCI America
Synonyms:
(S)-(2,2-Dimethyl-1,3-dioxolan-4-yl)methyl 4-methylbenzenesulfonate, (S)-(2,2-Dimethyl-1,3-dioxolan-4-yl)methyl 4-methylbenzenesulphonate, L-(+)-1,2-Isopropylideneglycerol 3-(p-Toluenesulphonate), L-(+)-1,2-Isopropylideneglycerol 3-(p-Toluenesulfonate)
CAS Number: 23735-43-5
MDL Number: MFCD00063234
Molecular Formula: C13H18O5S
Molecular Weight: 286.34
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 25
Specific rotation [a]20/D: 8 deg (C=2, DMF)
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Chiral Building Blocks
Other Product Names / Synonyms: L-(+)-1,2-Isopropylideneglycerol 3-(p-Toluenesulfonate)
Formula:
C₁₃H₁₈O₅S MW: 286.34 g/mol Flash Pt: 110 °C |
MDL Number:
MFCD00063234 CAS Number: 23735-43-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...