Ethyl diphenylphosphonoacetate ≥96.0%

Supplier: TCI America

Synonyms: Diphenylphosphonoacetic Acid Ethyl Ester, Ethyl 2-(diphenoxyphosphoryl)acetate

D2547-1G D2547-5G
TCD2547-001GEA 204.93 CAD
TCD2547-001G TCD2547-5G
Ethyl diphenylphosphonoacetate ≥96.0%
Ethyl diphenylphosphonoacetate

[Horner-Emmons Reagent]
CAS Number: 16139-79-0
MDL Number: MFCD01321165
Molecular Formula: C16H17O5P
Molecular Weight: 320.28
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 220
Specific Gravity (20/20): 1.22
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: Diphenylphosphonoacetic Acid Ethyl Ester

Formula: C₁₆H₁₇O₅P
MW: 320.28 g/mol
Boiling Pt: 220 °C
Density: 1.22 (20 °C)
MDL Number: MFCD01321165
CAS Number: 16139-79-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b695d30e345ff312066d0c51fe7b9e8c37f9bbef&vid=b695d30e345ff312066d0c51fe7b9e8c37f9bbef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR