Di-mu-hydroxo-bis[(N,N,N',N'-tetramethylethylenediamine)copper(II)] chloride ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Di-μ-hydroxo-bis[(N,N,N'.N'-tetramethylethylenediamine)copper(II)] chloride

D2542-25G D2542-5G
TCD2542-25GEA 255.96 CAD
TCD2542-25G TCD2542-005G
Di-mu-hydroxo-bis[(N,N,N',N'-tetramethylethylenediamine)copper(II)] chloride ≥97.0% (by titrimetric analysis)
Di-mu-hydroxo-bis[(N,N,N',N'-tetramethylethylenediamine)copper(II)] chloride

CAS Number: 30698-64-7
MDL Number: MFCD01321163
Molecular Formula: C12H34Cl2Cu2N4O2
Molecular Weight: 464.42
Purity/Analysis Method: >97.0% (T)
Form: Crystal

Formula: C₁₂H₃₄Cl₂Cu₂N₄O₂
MW: 464.42 g/mol
MDL Number: MFCD01321163
CAS Number: 30698-64-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b251c7368c3d2f940d5011239d5f52b8423abff3&vid=b251c7368c3d2f940d5011239d5f52b8423abff3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR