Difluoroacetaldehyde ethyl hemiacetal ≥90.0% (by NMR)

Supplier: TCI America

D2523-5G D2523-25G
TCD2523-005GEA 334.3 CAD
TCD2523-005G TCD2523-25G
Difluoroacetaldehyde ethyl hemiacetal ≥90.0% (by NMR)
Difluoroacetaldehyde ethyl hemiacetal

CAS Number: 148992-43-2
MDL Number: MFCD01321160
Molecular Formula: C4H8F2O2
Molecular Weight: 126.10
Purity/Analysis Method: >90.0% (NMR)
Form: Clear Liquid
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Ethoxy-2,2-difluoroethanol

Formula: C₄H₈F₂O₂
MDL Number: MFCD01321160
CAS Number: 148992-43-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ee0e4dc607305f04b6b119cd821cedd24b63a41&vid=2ee0e4dc607305f04b6b119cd821cedd24b63a41&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (NMR)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR