1,3-Di(2-pyridyl)-1,3-propanedione ≥98.0% (by GC, titration analysis)
Supplier: TCI America
D2512-5G
D2512-1G
TCD2512-005GEA
320.74
CAD
TCD2512-005G
TCD2512-1G
1,3-Di(2-pyridyl)-1,3-propanedione ≥98.0% (by GC, titration analysis)
1,3-Di(2-pyridyl)-1,3-propanedione
CAS Number: 10198-89-7
MDL Number: MFCD01321157
Molecular Formula: C13H10N2O2
Molecular Weight: 226.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 106
Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: Dipicolinoylmethane
Formula:
C₁₃H₁₀N₂O₂ Melting Pt: 106 °C |
MDL Number:
MFCD01321157 CAS Number: 10198-89-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...