Diphenyliodonium-2-carboxylate monohydrate ≥98.0%

Supplier: TCI America

Synonyms: (2-Carboxyphenyl)phenyliodonium Hydroxide Inner Salt

D2503-5G D2503-25G
TCD2503-005GEA 632 CAD
TCD2503-005G TCD2503-25G
Diphenyliodonium-2-carboxylate monohydrate ≥98.0%
Diphenyliodonium-2-carboxylate monohydrate

CAS Number: 96195-89-0
MDL Number: MFCD00150328
Molecular Formula: C13H9IO2
Molecular Weight: 324.12
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Hypervalent Iodine Compounds, Benzyne Precursors (Multi-component Reaction), Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: (2-Carboxyphenyl)phenyliodonium Hydroxide Inner Salt

Formula: C₁₃H₉IO₂
MW: 324.12 g/mol
MDL Number: MFCD00150328
CAS Number: 96195-89-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02e07714767e0955055dd6349c4d81d6b2b6e978&vid=02e07714767e0955055dd6349c4d81d6b2b6e978&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR