(2-Butoxyethoxy)acetic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
3,6-Dioxadecanoic Acid
D2491-5G
D2491-25G
TCD2491-005GEA
188.02
CAD
TCD2491-005G
TCD2491-025G
(2-Butoxyethoxy)acetic acid ≥98.0% (by GC, titration analysis)
(2-Butoxyethoxy)acetic acid
CAS Number: 82941-26-2
MDL Number: MFCD01105178
Molecular Formula: C8H16O4
Molecular Weight: 176.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 141
Specific Gravity (20/20): 1.05
Other Product Names / Synonyms: 3,6-Dioxadecanoic Acid
Formula:
C₈H₁₆O₄ MW: 176.21 g/mol Boiling Pt: 141 °C Density: 1.05 (20 °C) |
MDL Number:
MFCD01105178 CAS Number: 82941-26-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...