Diphenylphosphinic anhydride ≥98.0%
Supplier: TCI America
D2484-5G
D2484-1G
TCD2484-005GEA
1554.72
CAD
TCD2484-005G
TCD2484-001G
Diphenylphosphinic anhydride ≥98.0%
Diphenylphosphinic anhydride
CAS Number: 5849-36-5
MDL Number: MFCD01112176
Molecular Formula: C24H20O3P2
Molecular Weight: 418.37
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 146
Formula:
C₂₄H₂₀O₃P₂ Melting Pt: 146 °C |
MDL Number:
MFCD01112176 CAS Number: 5849-36-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...