3-Dimethylamino-2-methyl-2-propenal ≥97.0%

Supplier: TCI America

Synonyms: alpha-Methyl-beta-dimethylaminoacrolein

D2483-5G D2483-25G
TCD2483-005GEA 175.38 CAD
TCD2483-005G TCD2483-25G
3-Dimethylamino-2-methyl-2-propenal ≥97.0%
3-Dimethylamino-2-methyl-2-propenal

CAS Number: 19125-76-9
MDL Number: MFCD00075471
Molecular Formula: C6H11NO
Molecular Weight: 113.16
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 37


Other Product Names / Synonyms: alpha-Methyl-beta-dimethylaminoacrolein

Formula: C₆H₁₁NO
MW: 113.16 g/mol
Flash Pt: 110 °C
MDL Number: MFCD00075471
CAS Number: 19125-76-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bed821ffec74ff68ea83562dadf735d39a0132cf&vid=bed821ffec74ff68ea83562dadf735d39a0132cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR