Diacetyllauroyl glycerol (so called)

Supplier: TCI America

D2482-500ML D2482-25ML
TCD2482-500MLEA 149.47 CAD
TCD2482-500ML TCD2482-025ML
Diacetyllauroyl glycerol (so called)
Diacetyllauroyl glycerol

[Plasticizer]
MDL Number: MFCD00671531
Form: Clear Liquid
Color: Very Pale Yellow

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fed0696c8e84d8416e7229baa66592a60b7823ad&vid=021bd2e82d2f3ed8a025dea501e1f86b072d3cb7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR