4-(Dimethylamino)phenyldiphenylphosphine ≥95.0%

Supplier: TCI America

D2478-1G D2478-5G
TCD2478-001GEA 82.16 CAD
TCD2478-001G TCD2478-005G
4-(Dimethylamino)phenyldiphenylphosphine ≥95.0%
4-(Dimethylamino)phenyldiphenylphosphine

CAS Number: 739-58-2
MDL Number: MFCD00192068
Molecular Formula: C20H20NP
Molecular Weight: 305.36
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 152


Product Categories: Synthetic Organic Chemistry, Mitsunobu Reaction, Ligands
Other Product Names / Synonyms: 4-(Dimethylamino)triphenylphosphine, 4-(Diphenylphosphino)-N,N-dimethylaniline

Formula: C₂₀H₂₀NP
Melting Pt: 152 °C
MDL Number: MFCD00192068
CAS Number: 739-58-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fbbe060d586495933835ed6a16e0b103a3c29c46&vid=fbbe060d586495933835ed6a16e0b103a3c29c46&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR