6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride ≥98.0%

Supplier: TCI America

D2472-5G D2472-25G
TCD2472-005GEA 56.88 CAD
TCD2472-005G TCD2472-025G
6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride ≥98.0%
6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride

CAS Number: 2328-12-3
MDL Number: MFCD00012744
Molecular Formula: C11H15NO2
Molecular Weight: 229.70
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 248

Formula: C₁₁H₁₆ClNO₂
Melting Pt: 248 °C
MDL Number: MFCD00012744
CAS Number: 2328-12-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8123b1390e1a7481969d220145f1da5fa4ca041d&vid=8123b1390e1a7481969d220145f1da5fa4ca041d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR