Diphenyl-2-pyridylphosphine ≥97.0%
Supplier: TCI America
Synonyms:
2-(Diphenylphosphino)pyridine
D2471-1G
D2471-5G
TCD2471-001GEA
33.18
CAD
TCD2471-001G
TCD2471-5G
Diphenyl-2-pyridylphosphine ≥97.0%
Diphenyl-2-pyridylphosphine
CAS Number: 37943-90-1
MDL Number: MFCD00192108
Molecular Formula: C17H14NP
Molecular Weight: 263.28
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 85
Product Categories: Synthetic Organic Chemistry, Mitsunobu Reaction, Ligands, Materials Chemistry, Precise Polymerization Reagents
Other Product Names / Synonyms: 2-(Diphenylphosphino)pyridine
Formula:
C₁₇H₁₄NP MW: 263.28 g/mol Melting Pt: 85 °C |
MDL Number:
MFCD00192108 CAS Number: 37943-90-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...