Dodecafluorosuberic acid ≥97.0% (by titrimetric analysis)
Supplier: TCI America
D2465-25G
D2465-5G
TCD2465-025GEA
443.98
CAD
TCD2465-025G
TCD2465-005G
Dodecafluorosuberic acid ≥97.0% (by titrimetric analysis)
Dodecafluorosuberic acid
CAS Number: 678-45-5
MDL Number: MFCD00153146
Molecular Formula: C8H2F12O4
Molecular Weight: 390.08
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 143
Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: Dodecafluoro-1,8-octanedioic Acid, Perfluoro-1,8-octanedioic Acid, Perfluorosuberic Acid
Formula:
C₈H₂F₁₂O₄ Melting Pt: 143 °C |
MDL Number:
MFCD00153146 CAS Number: 678-45-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...