4,4'-Dinitrostilbene ≥98.0% (by HPLC)
Supplier: TCI America
D2464-25G
D2464-5G
TCD2464-25GEA
1540.5
CAD
TCD2464-25G
TCD2464-005G
4,4'-Dinitrostilbene ≥98.0% (by HPLC)
4,4'-Dinitrostilbene
CAS Number: 2501-02-2
MDL Number: MFCD00017043
Molecular Formula: C14H10N2O4
Molecular Weight: 270.24
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 295
Formula:
C₁₄H₁₀N₂O₄ Melting Pt: 295 °C |
MDL Number:
MFCD00017043 CAS Number: 2501-02-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...