Diethylene glycol bis(p-toluenesulfonate) ≥96.0%

Supplier: TCI America

Synonyms: Oxybis(ethane-2,1-diyl) bis(4-methylbenzenesulphonate), Bis[2-(p-toluenesulfonyloxy)ethyl] Ether, Diethylene Glycol Ditosylate, Bis(2-tosyloxyethyl) Ether, Oxybis(ethane-2,1-diyl) bis(4-methylbenzenesulfonate), Bis[2-(p-toluenesulphonyloxy)ethyl] Ether

D2457-25G D2457-5G
TCD2457-025GEA 216.46 CAD
TCD2457-025G TCD2457-5G
Diethylene glycol bis(p-toluenesulfonate) ≥96.0%
Diethylene glycol bis(p-toluenesulfonate)

CAS Number: 7460-82-4
MDL Number: MFCD00026001
Molecular Formula: C18H22O7S2
Molecular Weight: 414.49
Purity/Analysis Method: >96.0% (HPLC,T)
Form: Crystal
Melting point (°C): 87


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: Bis[2-(p-toluenesulfonyloxy)ethyl] Ether, Bis(2-tosyloxyethyl) Ether, Diethylene Glycol Ditosylate

Formula: C₁₈H₂₂O₇S₂
MW: 414.49 g/mol
Melting Pt: 87 °C
MDL Number: MFCD00026001
CAS Number: 7460-82-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3576e5b5189c9a2b709e43e8aabd2b9a70f8a25&vid=f3576e5b5189c9a2b709e43e8aabd2b9a70f8a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR