Dimethyl-2,5-dioxahexanedioate ≥98.0%
Supplier: TCI America
D2455-25G
D2455-5G
TCD2455-025GEA
414.67
CAD
TCD2455-025G
TCD2455-5G
Dimethyl-2,5-dioxahexanedioate ≥98.0%
Dimethyl-2,5-dioxahexanedioate
CAS Number: 88754-66-9
MDL Number: MFCD00085447
Molecular Formula: C6H10O6
Molecular Weight: 178.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 220
Specific Gravity (20/20): 1.24
Other Product Names / Synonyms: 2,5-Dioxahexanedioic Acid Dimethyl Ester
Formula:
C₆H₁₀O₆ Boiling Pt: 220 °C Density: 1.24 (20 °C) |
MDL Number:
MFCD00085447 CAS Number: 88754-66-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...