2-Acetoxybenzonitrile ≥97.0%

Supplier: TCI America

A1118-25G A1118-5G
TCA1118-025GEA 185.18 CAD
TCA1118-025G TCA1118-5G
2-Acetoxybenzonitrile ≥97.0%
2-Acetoxybenzonitrile

CAS Number: 5715-02-6
MDL Number: MFCD00017360
Molecular Formula: C9H7NO2
Molecular Weight: 161.16
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 254
Specific Gravity (20/20): 1.14


Other Product Names / Synonyms: Acetic Acid 2-Cyanophenyl Ester, 2-Cyanophenyl Acetate

Formula: C₉H₇NO₂
Boiling Pt: 254 °C
Density: 1.14 (20 °C)
MDL Number: MFCD00017360
CAS Number: 5715-02-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d05cdc2a79e5604e8ceaa432c27984b57f7d201&vid=1d05cdc2a79e5604e8ceaa432c27984b57f7d201&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR