Diethyl-3,3-diethoxypropane-1,1-dicarboxylate ≥96.0%
Supplier: TCI America
Synonyms:
3,3-Diethoxypropane-1,1-dicarboxylic Acid Diethyl Ester, Diethyl 3,3-Diethoxypropane-1,1-dicarboxylate
D2438-5G
D2438-1G
TCD2438-5GEA
165.27
CAD
TCD2438-5G
TCD2438-001G
Diethyl-3,3-diethoxypropane-1,1-dicarboxylate ≥96.0%
Diethyl-3,3-diethoxypropane-1,1-dicarboxylate
CAS Number: 21339-47-9
MDL Number: MFCD00671525
Molecular Formula: C13H24O6
Molecular Weight: 276.33
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.02
Other Product Names / Synonyms: 3,3-Diethoxypropane-1,1-dicarboxylic Acid Diethyl Ester
Formula:
C₁₃H₂₄O₆ MW: 276.33 g/mol Density: 1.02 (20 °C) |
MDL Number:
MFCD00671525 CAS Number: 21339-47-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...