Diethyl (2,2-diethoxyethyl)phosphonate ≥96.0%

Supplier: TCI America

D2423-25G D2423-5G
TCD2423-025GEA 447.93 CAD
TCD2423-025G TCD2423-005G
Diethyl (2,2-diethoxyethyl)phosphonate ≥96.0%
Diethyl (2,2-diethoxyethyl)phosphonate

CAS Number: 7598-61-0
MDL Number: MFCD00009236
Molecular Formula: C10H23O5P
Molecular Weight: 254.26
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 130
Flash Point (°C): 110
Specific Gravity (20/20): 1.05
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: 2,2-Diethoxyethylphosphonic Acid Diethyl Ester, Diethylphosphonoacetaldehyde Diethyl Acetal

Formula: C₁₀H₂₃O₅P
Boiling Pt: 130 °C
Density: 1.05 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00009236
CAS Number: 7598-61-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a05c5cabc1d9d6eb3f0e2b13379d12d4c689786&vid=7a05c5cabc1d9d6eb3f0e2b13379d12d4c689786&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR