2,3-Dichloro-5,8-dihydroxy-1,4-naphthoquinone ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2,3-Dichloro-5,8-dihydroxynaphthalene-1,4-dione

D2421-1G D2421-5G
TCD2421-1GEA 233.84 CAD
TCD2421-1G TCD2421-005G
2,3-Dichloro-5,8-dihydroxy-1,4-naphthoquinone ≥97.0% (by HPLC, titration analysis)
2,3-Dichloro-5,8-dihydroxy-1,4-naphthoquinone

CAS Number: 14918-69-5
MDL Number: MFCD00075261
Molecular Formula: C10H4Cl2O4
Molecular Weight: 259.04
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 201

Formula: C₁₀H₄Cl₂O₄
MW: 259.04 g/mol
Melting Pt: 201 °C
MDL Number: MFCD00075261
CAS Number: 14918-69-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0b0c764947a4b976edfec6b477207604f68ee34&vid=f0b0c764947a4b976edfec6b477207604f68ee34&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR