3,5-Diisopropylpyrazole ≥98.0%

Supplier: TCI America

Synonyms: 3,5-Diisopropyl-1H-pyrazole

D2407-25G D2407-5G
TCD2407-25GEA 995.08 CAD
TCD2407-25G TCD2407-005G
3,5-Diisopropylpyrazole ≥98.0%
3,5-Diisopropylpyrazole

CAS Number: 17536-00-4
MDL Number: MFCD00671516
Molecular Formula: C9H16N2
Molecular Weight: 152.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 88

Formula: C₉H₁₆N₂
Melting Pt: 88 °C
MDL Number: MFCD00671516
CAS Number: 17536-00-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cade330f08a8a43285f46fea2493c483a3974510&vid=cade330f08a8a43285f46fea2493c483a3974510&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR