5,6-Dichloro-3-pyridinemethanol ≥96.0%
Supplier: TCI America
Synonyms:
(5,6-Dichloropyridin-3-yl)methanol, 2,3-Dichloro-5-hydroxymethylpyridine, 5,6-Dichloronicotinyl Alcohol
D2402-1G
D2402-5G
TCD2402-1GEA
221.2
CAD
TCD2402-1G
TCD2402-005G
5,6-Dichloro-3-pyridinemethanol ≥96.0%
5,6-Dichloro-3-pyridinemethanol
CAS Number: 54127-30-9
MDL Number: MFCD00671515
Molecular Formula: C6H5Cl2NO
Molecular Weight: 178.01
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 76
Other Product Names / Synonyms: 2,3-Dichloro-5-hydroxymethylpyridine, 5,6-Dichloronicotinyl Alcohol
Formula:
C₆H₅Cl₂NO MW: 178.01 g/mol Melting Pt: 76 °C |
MDL Number:
MFCD00671515 CAS Number: 54127-30-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...